missing translation for 'onlineSavingsMsg'
Learn More
Learn More
xCT Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 26864-1-AP-20UL
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.00010kg
Description
SLC7A11 (also known as XCT) is a member of a hetromeric, sodium-independent, anionic amino acid transport system. It is highly specific for cysteine and glutamate. SLC7A11 is a mediator of Kaposi sarcoma-associated herpes virus fusion and entry into cells. Increased expression of this protein in primary gliomas is associated with neuronal cell death.Spécification
xCT | |
Polyclonal | |
Unconjugated | |
SLC7A11 | |
Amino acid transport system xc, CCBR1, Cystine/glutamate transporter, SLC7A11, xCT | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23657, 26570 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9UPY5, Q9WTR6 | |
SLC7A11 | |
xCT Fusion Protein Ag25431 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |