Learn More
Waters Corporation XBridge Prep Amide Column
XBridge BEH Amide columns retain extremely polar compounds, including carbohydrates and sugars.
Marque: Waters Corporation 186006605
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
- XBridge Amide columns offer you a chemically stable, trifunctionally bonded amide phase that is stable from pH 2 to 11 to enable the separation of polar analytes spanning a wide range of polarity, structural moiety and pKa.
Spécification
19 mm | |
6000 psi (415 Bar) | |
5 μm | |
1/Ea. | |
No |
150 mm | |
Sphérique | |
130 Å | |
L68 | |
Colonne |