Learn More
Waters Corporation XBridge Prep Amide Column
XBridge BEH Amide columns retain extremely polar compounds, including carbohydrates and sugars.
Marque:  Waters Corporation 186006610
Encompass
Code produit. 17123802
  • 11595.00€ / 1 pièce
Expédition estimée: 02-01-2025
pour voir le stock disponible 
Découvrez plus d'offres spéciales
Description
  • XBridge Amide columns offer you a chemically stable, trifunctionally bonded amide phase that is stable from pH 2 to 11 to enable the separation of polar analytes spanning a wide range of polarity, structural moiety and pKa.
Spécification
Spécification
30 mm
6000 psi (415 Bar)
5 μm
1/Ea.
No
150 mm
Sphérique
130 Å
L68
Colonne