Learn More
WNK1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 28357-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The WNK1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human WNK1. This antibody recognizes human antigen. The WNK1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Spécification
| WNK1 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 65125 | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| 367 μg/mL | |
| Rabbit | |
| Primary | |
| Human |