Learn More
Tocris Bioscience™ Vilazodone hydrochloride
Low molecular weight, organic compound that may help regulate biological process.
Marque: Tocris Bioscience™ 6202/50
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Vilazodone hydrochloride is a potent 5-HT1A partial agonist and serotonin-selective reuptake inhibitor (SSRI) (IC50 values are 0.2 and 0.5 nM respectively). Antidepressant.Specifications
Agonist | |
163521-08-2 | |
477.99g/mol | |
5-HT1A Receptor Antagonists |
5-HT1A | |
C26H27N5O2.HCl | |
Store at -20°C | |
50 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Usage exclusivement réservé à la recherche
Spot an opportunity for improvement?Share a Content Correction