Learn More
USP4 Mouse anti-Human, Mouse, Clone: 1G11G8, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 66822-1-IG-20UL
Description
The protein encoded by this gene is a protease that deubiquitinates target proteins such as ADORA2A and TRIM21. The encoded protein shuttles between the nucleus and cytoplasm and is involved in maintaining operational fidelity in the endoplasmic reticulum. Three transcript variants encoding different isoforms have been found for this gene.Spécification
USP4 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P35123, Q13107 | |
USP4 | |
USP4 Fusion Protein Ag28297 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG2b |
Western Blot | |
1G11G8 | |
Unconjugated | |
USP4 | |
Deubiquitinating enzyme 4, Ubiquitin thiolesterase 4, UNP, Unph, USP4 | |
Mouse | |
Protein A | |
RUO | |
22258, 7375 | |
-20°C | |
Liquid |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.