Learn More
Melford TEMED
Tertiary amine used to catalyze formation of free radicals from ammonium persulfate and riboflavin.
Marque:  Melford T18000-0.025
Code produit. 30018829
  • 41.10€ / 25mL
Découvrez plus d'offres spéciales
Description
Description
Tertiary amine used to catalyze formation of free radicals from ammonium persulfate and riboflavin.
Tertiary amine used to catalyze formation of free radicals from ammonium persulfate and riboflavin. Suitable for electrophoresis and tested as a catalyst for polyacrylamide gel electrophoresis.
Spécification
Spécification
TEMED
110-18-9
25 mL
C6H16N2