Learn More
Melford TEMED
Tertiary amine used to catalyze formation of free radicals from ammonium persulfate and riboflavin.
Marque: Melford T18000-0.025
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Tertiary amine used to catalyze formation of free radicals from ammonium persulfate and riboflavin.
Tertiary amine used to catalyze formation of free radicals from ammonium persulfate and riboflavin. Suitable for electrophoresis and tested as a catalyst for polyacrylamide gel electrophoresis.Spécification
TEMED | |
110-18-9 | |
25 mL | |
C6H16N2 |