Learn More
Thermo Scientific™ TDE Series -86°C Ultra-Low Temperature Chest Freezers, General Purpose
Part of the Standard Performance (STP) family, Thermo Scientific™ TDE Series ultra-low temperature chest freezers, powered by H-drive, feature three sizes, maximizing storage capacity from 3,645 up to 21,600 2mL vials. The TDE Series is a sustainable and reliable solution with ultimate sample security and operational savings for every lab.
11959.50€ - 16783.50€
Spécification
Gamme de températures | -50°C à -86°C |
---|---|
Modèle de porte | Solide horizontal |
No. of Doors | 1 |
Matériau de la hotte | Acier laminé à froid |
Intérieur | Acier inoxydable |
Code produit | Marque | Capacité | Capacité (métrique) | Certifications/Conformité | Tension | Prix | Quantité et disponibilité | ||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Capacité | Capacité (métrique) | Certifications/Conformité | Tension | Prix | Quantité et disponibilité | ||
17734579
|
Thermo Scientific™
TDEC06386FV |
Boîtes standard de 2 pouces (63) | 85 L | CE, ISO 13485, RoHS, REACH, EPA SNAP, UKCA, Prop 65 | 230 V |
11959.50€
1 pièce |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||
17744579
|
Thermo Scientific™
TDEC25286FV |
Boîtes standard de 2 pouces (252) | 360 L | CE, ISO 13485, RoHS, REACH, EPA SNAP, UKCA, Prop 65 | 230 V |
14874.00€
1 pièce |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||
17754579
|
Thermo Scientific™
TDEC39686FV |
Boîtes standard de 2 pouces (396) | 566 L | CE, ISO 13485, RoHS, REACH, EPA SNAP, UKCA, Prop 65 | 230 V |
16783.50€
1 pièce |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||
Description
Thermo Scientific™ TDE Series ultra-low temperature chest freezers, part of the Standard Performance (STP) family of freezers powered by H-drive, feature three sizes, maximizing storage capacity from 3,645 up to 21,600 2mL vials. The workhorse of the busy lab, the TDE Series sets the standard for performance and reliable sample storage in heavy use situations. A sustainable solution with ultimate sample security and operational savings for every lab. From our first ULT freezer in 1940 to today, the combined experience of REVCO, Forma Scientific, Jewett, Harris, and other families of products continues to drive our ability to deliver industry-leading innovations that help accelerate science and health care.
Performance
- Powered by H-drive's hydrocarbon technology, these systems save up to 25% in energy usage and heat rejection over conventional refrigerant ultra-low freezers1
- Designed for daily usage, recovers cabinet temperature 20% faster after opening the door.2
- A quiet sound output below 55 dBA, enables the freezer to come out of the hallway and into the lab
- Meets +/-10°C validation protocols at -80°C with strong peak variation and steady-state temperature variation
- Natural hydrocarbon refrigerants reduce system operating stresses promoting outstanding reliability and worry-free sample protection
User Friendly
- The H-drive Information Center (HIC) combines easy to read temperature with simple capacitive touch buttons in a 5.6 in. display
- Three sizes (3, 13, and 20 cu. ft.) to accommodate a variety of storage needs and lab footprint specifications
- Improved customer convenience with new ergonomically designed handle, high contrast easy-to-read display, and convenient additional workspace.
- Compatible with most racking systems including a full range of Thermo Scientific™ racks for boxes, microplates, Thermo Scientific™ Matrix™ and Nunc CryoBank™ tubes
- Security features including alarm settings and key-lock door
Sustainability
- Lower HVAC costs as a result of less heat emission into the environment
- Natural refrigerants and Polyurethane foam with non-HFC blowing agent having zero GWP, making the Series compliant with the U.S. Environmental Protection Agency's Significant New Alternatives Policy (SNAP), the European Union's F-Gas and other sustainability standards
- Upon receiving the unit, we advise you to properly dispose of any packaging material, used to secure the unit during the shipping process. Please refer to local recycling codes and dispose of any materials accordingly. For information on waste disposal sites, please refer to your local waste provider.
Get Connected
The Thermo Scientific™ DeviceLink™ Connect remote monitoring solution helps provide peace of mind by enabling you to keep track of your equipment anytime, anywhere. This system supports independent sensors to monitor your instrument, but it can also be connected via serial port directly to the instrument, harnessing the on-board telemetry data, alarms, and events.
Spécification
-50°C à -86°C | |
1 | |
Acier inoxydable | |
Interface utilisateur tactile capacitive | |
Un orifice de port | |
Levage d’une seule main | |
Alarme, enregistreur | |
Identification unique | |
Compatibilité avec Smart View | |
Compresseur H-Drive |
Solide horizontal | |
Acier laminé à froid | |
Acier peint | |
Sauvegarde LN2 en option, sauvegarde CO2, enregistreur graphique | |
Manuel | |
Non | |
RS485, 4-20 Ma, contacts sans potentiel, USB | |
En option (Connexion Device Link) | |
Unique | |
Mélange R290 / R170+R290 |