missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRBD1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 24931-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
SRBD1 gene ontology annotations related to this gene include nucleobase-containing compound metabolic process; translation.Spécification
SRBD1 | |
Polyclonal | |
Unconjugated | |
SRBD1 | |
S1 RNA binding domain 1, SRBD1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55133, 78586 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q497V5, Q8N5C6 | |
SRBD1 | |
SRBD1 Fusion Protein Ag21661 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |