Learn More
Gilac™ Square Bucket with Watertight Lid, 12 L
Square 12 L Bucket with Watertight Lid fit for food contact with watertight lid and pouring spout.
Marque:  Gilac™ G615710
Code produit. 17998431
  • 43.60€ / 1 pièce
Expédition estimée: 15-04-2025
pour voir le stock disponible 
Découvrez plus d'offres spéciales
Description
  • Bucket fit for food contact with watertight lid and pouring spout.
  • Airtight lid keeps flavours in and odours out.
  • Nesting when empty, no suction and stacking when lid is closed.
  • Enables liquid preparations to be stored and transported.
  • Graduation in litres.
  • Discharge area that prevents water retention when being cleaned and anti-wear skirt that lifts the bucket off the ground.
  • HDPE body and LDPE lid.
  • Stainless steel handle.
  • External dimensions include handle.
Spécification
Spécification
Bucket, Handle, and Lid
12 L
Square
Documents
One moment while we fetch your results.
Nom du produit
Gilac™ Square Bucket with Watertight Lid, 12 L > Couleur : Bleu