missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPOCD1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 22243-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Spécification
SPOCD1 | |
Polyclonal | |
Unconjugated | |
SPOCD1 | |
RP11 84A19.1, SPOC domain containing 1, SPOCD1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
90853 | |
-20°C | |
Liquid |
Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6ZMY3 | |
SPOCD1 | |
SPOCD1 Fusion Protein Ag17734 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |