Learn More
BUCHI Rotavapor Vapor Duct with Combi-Clip
Vapor duct with Combi-Clip for use with connecting the evaporating flask to the Rotavapor.
Marque: BUCHI 11062186
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
- Combi-Clip is included for fastening the evaporating flask
Spécification
Borosilicate Glass 3.3 | |
Glass Assembly V, C, S, E, CR, BY, HP | |
Combi-Clip |
Vapor Duct | |
Rotavapor R-300 | |
29/32 |