Learn More
BUCHI Rotavapor R-100 Vertical Condenser Glass Assembly
Vertical Condenser Glass Assembly for Rotavapor R-100, 1500 cm2. For standard distillations, used with recirculating chiller or tap water. Includes a 1 liter receiving flask, required tubing, and a ball joint clamp.
Marque: BUCHI 11057057
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
- Includes receiving flask (1 L), tubing, and a ball joint clamp
- Does NOT include an evaporating flask, vacuum seal, vapor duct, or condenser holder
- For use with Rotavapor R-100, R-3
Spécification
Borosilicate Glass (3.3) | |
Rotavapor R-100, R-3 | |
P+G Safety Coating |
Vertical Condensor | |
Receiving Flask (1 L), Tubing, and a Ball Joint Clamp | |
1000 mL |