missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RLN2 Protein

A cDNA sequence encoding the RLN2 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP13322-HIS-25ug

Code nomenclature Nacres: NA.28

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15983199

  • 203.00€ / 25µg
Expédition estimée: 09-10-2024
pour voir le stock disponible 

Description

Description

Spécification

Spécification

RLN2 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSDSWMEEV IKLCGRELVR AQIAICGMST WSKRSLSQED APQTPRPVAE IVPSFINKDT ETINMMSEFV ANLPQELKLT LSEMQPALPQ LQQHVPVLKD SSLLFEEFKK LIRNRQSEAA DSSPSELKYL GLDTHSRKKR QLYSALANKC CHVGCTKRSL ARFC
Greater than 85.0% as determined by SDS-PAGE.
25 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
RLN2 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol.
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions