missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PREP Protein
A cDNA sequence encoding the PREP was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13136-100ug
Code nomenclature Nacres: NA.28
Informations supplémentaires : Poids : 0.01000kg
Spécification
5550 | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMLSLQYP DVYRDETAVQ DYHGHKICDP YAWLEDPDSE QTKAFVEAQN KITVPFLEQC PIRGLYKERM TELYDYPKYS CHFKKGKRYF YFYNTGLQNQ RVLYVQDSLE GEARVFLDPN ILSDDGTVAL RGYAFSEDGE YFAYGLSASG SDWVTIKFMK VDGAKELPDV LERVKFSCMA WTHDGKGMFY NSYPQQDGKS DGTETSTNLH QKLYYHVLGT DQSEDILCAE FPDEPKWMGG AELSDDGRYV LLSIREGCDP VNRLWYCDLQ QESSGIAGIL KWVKLIDNFE GEYDYVTNEG TVFTFKTNRQ SPNYRVINID FRDPEESKWK VLVPEHEKDV LEWIACVRSN FLVLCYLHDV KNILQLHDLT TGALLKTFPL DVGSIVGYSG QKKDTEIFYQ FTSFLSPGII YHCDLTKEEL EPRVFREVTV KGIDASDYQT VQIFYPSKDG TKIPMFIVHK KGIKLDGSHP AFLYGYGGFN ISITPNYSVS RLIFVRHMGG ILAVANIRGG GEYGETWHKG GILANKQNCF DDFQCAAEYL IKEGYTSPKR LTINGGSNGG LLVAACANQR PDLFGCVIAQ VGVMDMLKFH KYTIGHAWTT DYGCSDSKQH FEWLVKYSPL HNVKLPEADD IQYPSMLLLT ADHDDRVVPL HSLKFIATLQ YIVGRSRKQS NPLLIHVDTK AGHGAGKPTA KVIEEVSDMF AFIARCLNVD WIP | |
Greater than 90.0% as determined by SDS-PAGE. |
PREP Protein | |
Research Use Only | |
PREP | |
Human | |
His | |
PREP protein solution (0.25 mg/ml) containing PBS buffer (pH 7.4), 30% glycerol and 1mM DTT. |