Learn More
enQuireBio™ Recombinant Human POLD4 Protein
A cDNA sequence encoding the POLD4 was constructed and used to recombinantly synthesize the protein.
Marque:  enQuireBio™ QP13092-10ug
Code produit. 15982479
  • 203.00€ / 10µg
Découvrez plus d'offres spéciales
Spécification
57804
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMGRKRLI TDSYPVVKRR EGPAGHSKGE LAPELGEEPQ PRDEEEAELE LLRQFDLAWQ YGPCTGITRL QRWCRAKQMG LEPPPEVWQV LKTHPGDPRF QCSLWHLYPL
Greater than 90.0% as determined by SDS-PAGE.
POLD4 Protein
Research Use Only
POLD4
Human
His
The POLD4 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 40% glycerol and 2mM DTT.