missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PLA2G7 Protein
A cDNA sequence encoding the PLA2G7 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13074-HIS-HEK-20ug
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Spécification
PLA2G7 Protein | |
Research Use Only | |
Recombinant Protein | |
Human (HEK293) | |
MVPPKLHVLF CLCGCLAVVY PFDWQYINPV AHMKSSAWVN KIQVLMAAAS FGQTKIPRGN GPYSVGCTDL MFDHTNKGTF LRLYYPSQDN DRLDTLWIPN KEYFWGLSKF LGTHWLMGNI LRLLFGSMTT PANWNSPLRP GEKYPLVVFS HGLGAFRTLY SAIGIDLASH GFIVAAVEHR DRSASATYYF KDQSAAEIGD KSWLYLRTLK QEEETHIRNE QVRQRAKECS QALSLILDID HGKPVKNALD LKFDMEQLKD SIDREKIAVI GHSFGGATVI QTLSEDQRFR CGIALDAWMF PLGDEVYSRI PQPLFFINSE YFQYPANIIK MKKCYSPDKE RKMITIRGSV HQNFADFTFA TGKIIGHMLK LKGDIDSNVA IDLSNKASLA FLQKHLGLHK DFDQWDCLIE GDDENLIPGT NINTTNQHIM LQNSSGIEKY N | |
Greater than 95.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
PLA2G7 protein is supplied in 20mM Tris-HCl pH 8.0, 1mM EDTA and 50% glycerol. |