missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PDAP1 Protein
A cDNA sequence encoding the PDAP1 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12984-5ug
Code nomenclature Nacres: NA.28
Informations supplémentaires : Poids : 0.01000kg
Spécification
PDAP1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MPKGGRKGGH KGRARQYTSP EEIDAQLQAE KQKAREEEEQ KEGGDGAAGD PKKEKKSLDS DESEDEEDDY QQKRKGVEGL IDIENPNRVA QTTKKVTQLD LDGPKELSRR EREEIEKQKA KERYMKMHLA GKTEQAKADL ARLAIIRKQR EEAARKKEEE RKAKDDATLS GKRMQSLSLN KLEHHHHHH | |
Greater than 90.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PDAP1 solution (1 mg/ml) contains 20mM Tris-HCl pH 8.0, 2mM DTT, 0.1M NaCl, 0.1mM PMSF and 20% glycerol. |