missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NBL1 Protein
A cDNA sequence encoding the NBL1 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12797-1mg
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Spécification
NBL1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSAPPPINK LALFPDKSAW CEAKNITQIV GHSGCEAKSI QNRACLGQCF SYSVPNTFPQ STESLVHCDS CMPAQSMWEI VTLECPGHEE VPRVDKLVEK ILHCSCQACG KEPSHEGLSV YVQGEDGPGS QPGTHPHPHP HPHPGGQTPE PEDPPGAPHT EEEGAED | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
NBL1 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 10% glycerol and 1mM DTT. |