missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IF Protein
A cDNA sequence encoding the IF was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12371-1mg
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Spécification
IF Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
MAWFALYLLS LLWATAGTST QTQSSCSVPS AQEPLVNGIQ VLMENSVTSS AYPNPSILIA MNLAGAYNLK AQKLLTYQLM SSDNNDLTIG QLGLTIMALT SSCRDPGDKV SILQRQMENW APSSPNAEAS AFYGPSLAIL ALCQKNSEAT LPIAVRFAKT LLANSSPFNV DTGAMATLAL TCMYNKIPVG SEEGYRSLFG QVLKDIVEKI SMKIKDNGII GDIYSTGLAM QALSVTPEPS KKEWNCKKTT DMILNEIKQG KFHNPMSIAQ ILPSLKGKTY LDVPQVTCSP DHEVQPTLPS NPGPGPTSAS NITVIYTINN QLRGVELLFN ETINVSVKSG SVLLVVLEEA QRKNPMFKFE TTMTSWGLVV SSINNIAENV NHKTYWQFLS GVTPLNEGVA DYIPFNHEHI TANFTQY | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The protein solution (0.7 mg/ml) contains 20mM HEPES pH 8.0, 100mM NaCl, 20% Glycerol. |