missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD40 Protein
A cDNA sequence encoding the CD40 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11333-2ug
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Spécification
958 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSEPPTACR EKQYLINSQC CSLCQPGQKL VSDCTEFTET ECLPCGESEF LDTWNRETHC HQHKYCDPNL GLRVQQKGTS ETDTICTCEE GWHCTSEACE SCVLHRSCSP GFGVKQIATG VSDTICEPCP VGFFSNVSSA FEKCHPWTSC ETKDLVVQQA GTNKTDVVCG PQDRLR | |
Greater than 90.0% as determined by SDS-PAGE. |
CD40 Protein | |
Research Use Only | |
CD40 | |
Human | |
His | |
CD40 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |