missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis B HBV core delta Protein
A cDNA sequence encoding the HBV core delta was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12131-500ug
Code nomenclature Nacres: NA.28
Informations supplémentaires : Poids : 0.01000kg
Spécification
Hepatitis B HBV core delta Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
7.5mM Phosphate buffer pH 7.2, 75mM NaCl, and 50% glycerol. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis B | |
Untagged | |
HBV Core Delta protein is >95% pure as determined by SDS-PAGE (coomassie staining). |