missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dengue Virus Dengue NS1 c Protein
A cDNA sequence encoding the Dengue NS1 c was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11642-1mg
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Spécification
Dengue NS1 c Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
LWSNGVLESE MVIPKNFAGP KSQHNNRPGY HTQTAGPWHL GKLEMDFDFC EGTTVVVTED CGNRGPSLRT TTASGKLITE WCCRSCTLPP LRYRGEDGCW YGMEIRPLKE KEENLVSSLV TAHHHHHH | |
Dengue Protein is >90% pure as determined by 10% PAGE (coomassie staining). |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Dengue Virus | |
Untagged | |
20mM Tris-HCl pH 8.0, 8M urea and 60mM NaCl. |