Learn More
Novus Biologicals™ Rat C-Reactive Protein/CRP ELISA Kit (Chemiluminescence)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Marque: Novus Biologicals™ NBP2-67252
Description
The Rat C-Reactive Protein/CRP ELISA Kit (Chemiluminescence) measures rat C-Reactive Protein/CRP in serum, plasma and other biological fluids.
Spécification
0.16-10 ng/mL | |
This kit recognizes Rat CRP in samples. No significant cross-reactivity or interference between Rat CRP and analogues was observed. | |
Biological Fluids, Plasma, Serum | |
1401 | |
Rat | |
Storage of components varies. See protocol for specific instructions. |
0.09 ng/mL | |
ELISA Kit (Chemiluminescence) | |
ELISA | |
CRP | |
1 kit | |
C-Reactive Protein/CRP |