Learn More
Propylamine, 98 %, pur, Thermo Scientific Chemicals
12.90€ - 331.00€
Identifiants chimiques
CAS | 107-10-8 |
---|---|
Numéro MDL | MFCD00008205 |
Clé InChI | WGYKZJWCGVVSQN-UHFFFAOYSA-N |
Synonyme | propylamine, 1-propanamine, 1-propylamine, propanamine, n-propylamine, 1-aminopropane, monopropylamine, mono-n-propylamine, propyl amine, n-propyl amine |
CID PubChem | 7852 |
ChEBI | CHEBI:39870 |
Nom IUPAC | propan-1-amine |
SMILES | CCCN |
Code produit | Marque | Quantité | Conditionnement | Prix | Quantité et disponibilité | ||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Conditionnement | Prix | Quantité et disponibilité | ||||
10500522
|
Thermo Scientific Acros
149310050 |
5 mL | Flacon en verre | N/A | |||||
10582731
|
Thermo Scientific Acros
149310010 |
1 L | Flacon en verre |
73.90€
1L |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15119203
|
Thermo Scientific Acros
149310051 |
5 L | Récipient en plastique | N/A | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
107-10-8 | |
WGYKZJWCGVVSQN-UHFFFAOYSA-N | |
7852 | |
propan-1-amine |
MFCD00008205 | |
propylamine, 1-propanamine, 1-propylamine, propanamine, n-propylamine, 1-aminopropane, monopropylamine, mono-n-propylamine, propyl amine, n-propyl amine | |
CHEBI:39870 | |
CCCN |
Spécification
Propylamine | |
107-10-8 | |
Incolore à jaune | |
48.0°C | |
Authentic | |
C3H9N | |
1.3870 to 1.3890 | |
0.719 | |
WGYKZJWCGVVSQN-UHFFFAOYSA-N | |
propan-1-amine | |
7852 | |
59.11 | |
Pures |
98% | |
-83.0°C | |
0.7190g/mL | |
−30°C | |
98% | |
CH3CH2CH2NH2 | |
MFCD00008205 | |
propylamine, 1-propanamine, 1-propylamine, propanamine, n-propylamine, 1-aminopropane, monopropylamine, mono-n-propylamine, propyl amine, n-propyl amine | |
CCCN | |
59.112 | |
CHEBI:39870 | |
98% | |
Liquid |
Sécurité et traitement
GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Highly flammable liquid and vapor.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
Recommended Storage : Réfrigérateur +4°C
RUO – Research Use Only