missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRDM10 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 23827-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
GPIP137 may regulate the transport and translation of mRNAs of proteins involved in synaptic plasticity in neurons and cell proliferation and migration in multiple cell types. It is expressed in the cytoplasm.Spécification
PRDM10 | |
Polyclonal | |
Unconjugated | |
PRDM10 | |
KIAA1231, PFM7, PR domain containing 10, PRDM10, TRIS, Tristanin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
382066, 56980 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3UTQ7, Q9NQV6 | |
Prdm10 | |
PRDM10 Fusion Protein Ag20605 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |