Learn More
PP 121
Dual kinase inhibitor; inhibits PI 3-K family kinases
Marque:  Tocris Bioscience 3894/5
82.07 EUR valable jusqu'au 2024-12-16
Utilisez le code promo "21630" pour bénéficier de cette offre.
Code produit. 18749206
  • 93.00€ / 5mg
Découvrez plus d'offres spéciales
Description
Description
PP 121 is a dual inhibitor of receptor tyrosine kinases (RTKs) (IC50 < 0.02 μM for Abl, Src, VEGFR-2 and PDGFR) and PI 3-K family kinases (IC50 < 0.06 μM for p110α, DNA-PK and mTOR). Exhibits no significant effect on receptor serine/threonine kinases (RSTKs). Blocks the proliferation of tumor cells by direct inhibition of PI 3-K, mTOR, Src and the VEGF receptor.
Spécification
Spécification
1-Cyclopentyl-3-(1H-pyrrolo[2,3-b]pyridin-5-yl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine
>99%
C17H17N7
1092788-83-4
5 mg
Observed MW: 319.36