Learn More
PKC epsilon Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP3-21224-25ul
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
PKC epsilon Polyclonal antibody specifically detects PKC epsilon in Human samples. It is validated for Immunofluorescence
Spécification
| PKC epsilon | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
| EC 2.7.11, EC 2.7.11.13, MGC125656, nPKC-epsilon, PKCEMGC125657, protein kinase C epsilon type, protein kinase C, epsilon | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: VDARGIAKVLADLGVTPDKITNSGQRRKKLIAGAESPQPASGSSPSEEDRSKSAPTSPCDQEIKELENNIRKALSFDNRGEEHRAASSPDGQLMSPGENGEVRQGQAKRLGLDEFNFIKVLG | |
| 25 μg | |
| Apoptosis, Cancer, Phospho Specific, Signal Transduction | |
| 5581 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |