missing translation for 'onlineSavingsMsg'
Learn More

Vitlab™ Opaque Volumetric Flask with Colored Screw Cap

Prepare chemical solutions with precision. Vitlab™ Opaque Volumetric Flasks with Colored Screw Cap are ideal for preparing and running chemical solutions in the laboratory. Flasks are UV-absorbing and able to store light-sensitive substances.

Marque:  Vitlab™ 674040

Code nomenclature Nacres: NB.43

Informations supplémentaires : Poids : 0.80000kg

 Afficher plus de versions de ce produit

Code produit. 10657645

Cet article est actuellement indisponible ou discontinu. Veuillez consulter la page produit pour de possible alternative dans l’onglet


Description

Description

  • UV-absobring yet highly transparentr
  • Thermal stress up to 121 °C does not permanently exceed the tolerance limit
  • To preserve markings, cleaning at no higher than 60 °C is recommended 
Spécification

Spécification

250 mL
235 mm
GL 25
PMP
121°C
0.15 mL
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions