Learn More
Kolmi Hopen™ Op-Air™ Type IIR Splash-resistant Medical Mask in Flowpack
Splash-resistant medical masks with earloops packaged conveniently in a flowpack for dental, medical, or industrial use.
Marque: Kolmi Hopen™ M35111-32-BULK
Informations supplémentaires : Poids : 5.22500kg
Description
- Protection against human liquids, bacteria, particles, and viruses
- Anatomical folding reduces exposure to face
- Inner layer designed to wick away moisture and sweat
- Convenient, hygienic, easy-to-use packaging
Spécification
Blue | |
Polypropylene | |
Pleated | |
Yes | |
Yes |
Earloops | |
175 x 90 mm | |
(EU) 2017/745, EN 14683:2019+AC:2019 | |
Medical | |
600 Pk. |