Learn More
Kolmi Hopen™ Op-Air™ Type IIR Splash-resistant Medical Mask in Flowpack
Splash-resistant medical masks with earloops packaged conveniently in a flowpack for dental, medical, or industrial use.
Marque:  Kolmi Hopen™ M35111-32
Informations supplémentaires : Poids : 2.37500kg
Code produit. 17750209
  • 2.50€ / 25 pièces
En stock
Description
  • Protection against human liquids, bacteria, particles, and viruses
  • Anatomical folding reduces exposure to face
  • Inner layer designed to wick away moisture and sweat
  • Convenient, hygienic, easy-to-use packaging
Spécification
Spécification
Bleu
Polypropylène
Plissée
Oui
Oui
Contours d’oreille
175 x 90 mm
CE, (UE) 2017/745, EN 14683:2019+AC:2019
Médical
25 Paquets