Learn More
Kolmi Hopen™ Op-Air™ Type IIR Splash-resistant Medical Mask in Flowpack
Splash-resistant medical masks with earloops packaged conveniently in a flowpack for dental, medical, or industrial use.
Marque: Kolmi Hopen™ M35111-32
Informations supplémentaires : Poids : 2.37500kg
Description
- Protection against human liquids, bacteria, particles, and viruses
- Anatomical folding reduces exposure to face
- Inner layer designed to wick away moisture and sweat
- Convenient, hygienic, easy-to-use packaging
Spécification
Bleu | |
Polypropylène | |
Plissée | |
Oui | |
Oui |
Contours d’oreille | |
175 x 90 mm | |
CE, (UE) 2017/745, EN 14683:2019+AC:2019 | |
Médical | |
25 Paquets |