Learn More
Kolmi Hopen™ Op-Air™ One Type IIR Splash-resistant Medical Mask
These splash-resistant medical masks are designed for those with sensitive skin, providing less irritation against the face.
Marque: Kolmi Hopen™ M34301-30
Informations supplémentaires : Poids : 1.52000kg
Description
- Protects environment from droplets from wearer
- Provides protection against biological fluids
- Effective against bacteria, particles, and viruses
- Non-irritating inner layer
- Inner layer is specially designed to reduce moisture and wick away sweat
- Used by healthcare workers to reduce risk of infection or in the operating room
Spécification
Miel | |
Polypropylène | |
(UE) 2017/745, EN 14683:2019+AC:2019, ISO 9001, ISO 13485 | |
≥ 99% | |
Oui |
Attaches | |
Plissée | |
Oui | |
Médical | |
40 Paquets |