missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFC2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15573-1-AP-20UL
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.00010kg
Description
Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.Spécification
NDUFC2 | |
Polyclonal | |
Unconjugated | |
NDUFC2 | |
B14.5b, CI B14.5b, Complex I B14.5b, HLC 1, NADHDH2, NDUFC2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4718, 68197 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O95298, Q9CQ54 | |
NDUFC2 | |
NDUFC2 Fusion Protein Ag7919 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |