Learn More
N-Heptane, qualité spectrophotométrique, supérieur à 99 % n-Heptane 96 % min, Thermo Scientific Chemicals
87.80€ - 256.00€
Identifiants chimiques
CAS | 142-82-5 |
---|---|
Formule moléculaire | C7H16 |
Poids moléculaire (g/mol) | 100.21 |
Numéro MDL | MFCD00009544 |
Clé InChI | IMNFDUFMRHMDMM-UHFFFAOYSA-N |
Synonyme | n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish |
CID PubChem | 8900 |
ChEBI | CHEBI:43098 |
Nom IUPAC | heptane |
SMILES | CCCCCCC |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
11338377
|
Thermo Scientific Alfa Aesar
032441.K2 |
1 L |
87.80€
1L |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
11318377
|
Thermo Scientific Alfa Aesar
032441.M1 |
4 x 1 L |
256.00€
4L |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
11328377
|
Thermo Scientific Alfa Aesar
032441.K7 |
4 L | |||||||
Description
n-Heptane is used in paints and coatings, as the rubber cement solvent and GC analysis, HPLC, spectrophotometry and environmental testing. Used as a primary reference standard in determining the knock rating of carburetor fuels and solvent especially for vegetable oils, low temperature thermo- meters, calibrations, and polymerization reaction medium. The hydroisomerization of n-heptane to form dibranched and tribranched products useful for producing high octane gasoline. water/AOT/n-heptane microemulsions plays an essential role for the synthesis, size control, and passivation of CdS nanoparticles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
142-82-5 | |
100.21 | |
IMNFDUFMRHMDMM-UHFFFAOYSA-N | |
8900 | |
heptane |
C7H16 | |
MFCD00009544 | |
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish | |
CHEBI:43098 | |
CCCCCCC |
Specifications
n-Heptane | |
142-82-5 | |
Colorless liquid | |
97°C to 99°C | |
Petroleum-like | |
1.3877 | |
C7H16 | |
53 hPa (40mm Hg) at 20°C | |
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish | |
IMNFDUFMRHMDMM-UHFFFAOYSA-N | |
heptane | |
8900 | |
100.21 |
°C to 91°C | |
Incolore | |
0.684 | |
-4°C (24°F) | |
Liquide | |
>99% | |
MFCD00009544 | |
1730763 | |
Miscible with alcohol,chloroform and ether. Immiscible with water. | |
CCCCCCC | |
100.21 | |
CHEBI:43098 | |
Sprectrophotométrique |
Safety and Handling
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P331-P332+P313-P363-P370+P378q-P501c
H225-H304-H315-H319-H336-H373
missing translation for 'dotInformation' : DOT Class: 3; Packing Group: II
missing translation for 'einecsNumber' : 205-563-8
missing translation for 'rtecsNumber' : MI7700000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only