Learn More
MYH14 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 20716-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The antibody is specific to MYH14.
Cellular myosin that appears to play a role in cytokinesis, cell shape, and specialized functions such as secretion and capping.Spécification
MYH14 | |
Polyclonal | |
Unconjugated | |
MYH14 | |
MHC16, MYH14, myosin, Myosin 14, Myosin heavy chain 14, myosin, heavy chain 14, NMHC II C, non-muscle II c | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
308572, 71960, 79784 | |
-20°C | |
Liquid |
Western Blot, Immunofluorescence, Immunoprecipitation, Immunocytochemistry | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6URW6, Q7Z406 | |
Myh14 | |
Peptide | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |