Learn More
PAX6, Mouse, Clone: 1C8, Abnova™
Mouse monoclonal antibody raised against recombinant PAX6.
Marque: Abnova MAB10293.100uL
Description
This gene encodes paired box gene 6, one of many human homologs of the Drosophila melanogaster gene prd. In addition to the hallmark feature of this gene family, a conserved paired box domain, the encoded protein also contains a homeo box domain. Both domains are known to bind DNA, and function as regulators of gene transcription. This gene is expressed in the developing nervous system, and in developing eyes. Mutations in this gene are known to cause ocular disorders such as aniridia and Peter's anomaly. Alternatively spliced transcript variants encoding either the same or different isoform have been found for this gene. [provided by RefSeq]
Spécification
PAX6 | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Western Blot (1:500-1:2000) Flow cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. | |
PAX6 | |
PAX6 | |
Recombinant protein corresponding to human PAX6. | |
RUO | |
5080 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
ELISA, Flow Cytometry, Western Blot | |
1C8 | |
Mouse monoclonal antibody raised against recombinant PAX6. | |
In ascites (0.03% sodium azide) | |
AN/AN2/D11S812E/MGC17209/MGDA/WAGR | |
Mouse | |
100 μL | |
Primary | |
Human | |
Liquid |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.