Learn More
Phosphate de lithium, extra pur, Thermo Scientific Chemicals
63.20€ - 228.00€
Identifiants chimiques
CAS | 10377-52-3 |
---|---|
Formule moléculaire | Li3O4P |
Poids moléculaire (g/mol) | 115.79 |
Numéro MDL | MFCD00016187 |
Clé InChI | TWQULNDIKKJZPH-UHFFFAOYSA-K |
Synonyme | lithium phosphate, lithium orthophosphate, trilithium orthophosphate, trilithium phosphate, phosphoric acid, trilithium salt, unii-2qm4k05q74, lithiumphosphate, trilithotab phosphate, phosphoric acid, lithium salt 1:3, li3po4 |
CID PubChem | 165867 |
SMILES | [Li+].[Li+].[Li+].[O-]P([O-])([O-])=O |
Product Code | Brand | Quantité | Conditionnement | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantité | Conditionnement | Price | Quantity & Availability | ||||
10569413
|
Thermo Scientific Acros
389781000 |
100 g | Flacon en plastique |
63.20€
100g |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
10127053
|
Thermo Scientific Acros
389785000 |
500 g | Flacon en plastique |
228.00€
500g |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10377-52-3 | |
115.79 | |
TWQULNDIKKJZPH-UHFFFAOYSA-K | |
165867 |
Li3O4P | |
MFCD00016187 | |
lithium phosphate, lithium orthophosphate, trilithium orthophosphate, trilithium phosphate, phosphoric acid, trilithium salt, unii-2qm4k05q74, lithiumphosphate, trilithotab phosphate, phosphoric acid, lithium salt 1:3, li3po4 | |
[Li+].[Li+].[Li+].[O-]P([O-])([O-])=O |
Specifications
Lithium phosphate | |
10377-52-3 | |
Poudre cristalline | |
Li3PO4 | |
lithium phosphate, lithium orthophosphate, trilithium orthophosphate, trilithium phosphate, phosphoric acid, trilithium salt, unii-2qm4k05q74, lithiumphosphate, trilithotab phosphate, phosphoric acid, lithium salt 1:3, li3po4 | |
TWQULNDIKKJZPH-UHFFFAOYSA-K | |
Trilithium ; phosphate | |
165867 | |
≥95.0% |
837°C | |
Blanc | |
Li3O4P | |
MFCD00016187 | |
Solubility in water: 0.4g/L (20°C) | |
[Li+].[Li+].[Li+].[O-]P([O-])([O-])=O | |
115.79 | |
115.79 | |
Extra pur |
Safety and Handling
Déclaration GHS H :
Provoque une irritation cutanée.
Nocif en cas d’ingestion.
Peut irriter les voies respiratoires.
Provoque une grave irritation oculaire.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Mention d’avertissement GHS :Avertissement
missing translation for 'einecsNumber' : 233-823-
- Lithium phosphate
Signal Word
- Attention
Hazard Category
- Toxicité aiguë Catégorie 4
- Lésion oculaire grave/irritation oculaire Catégorie 2
- Corrosion cutanée/irritation cutanée Catégorie 2
- Toxicité spécifique pour certains organes cibles Catégorie 3
Hazard Statement
- H315-Provoque une irritation cutanée.
- H319-Provoque une sévère irritation des yeux.
- H335-Peut irriter les voies respiratoires.
- H302-Nocif en cas d'ingestion.
Precautionary Statement
- P261-Éviter de respirer les poussières/fumées/gaz/brouillards/vapeurs/ aérosols.
- P280-Porter des gants de protection/des vêtements de protection/un équipement de protection des yeux/ du visage.
- P301+P312-EN CAS D'INGESTION: appeler un CENTRE ANTIPOISON/médecin / en cas de malaise.
- P302+P352-EN CAS DE CONTACT AVEC LA PEAU: Laver abondamment à l'eau/.
- P305+P351+P338-EN CAS DE CONTACT AVEC LES YEUX: rincer avec précaution à l'eau pendant plusieurs minutes. Enlever les lentilles de contact si la victime en porte et si elles peuvent être facilement enlevées. Continuer à rincer.
RUO – Research Use Only