missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ki67/MKI67 Antibody (1297A) - BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody has been used in 2 publications
185.00€ - 442.00€
Spécification
Antigène | Ki67/MKI67 |
---|---|
Clone | 1297A |
Concentration | 1.0 mg/mL |
Dilution | Flow Cytometry 1 μg/mL, Immunohistochemistry 3-15 μg/mL, Immunocytochemistry/Immunofluorescence 1-10 μg/mL, Immunohistochemistry-Paraffin 3-15 μg/mL, Flow (Intracellular) 1 μg/mL, Knockout Validated |
Applications | Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
18219720
|
Bio-Techne
NBP2-54791SS |
0.025 mg |
185.00€
25µg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
18009073
|
Bio-Techne
NBP2-54791 |
0.1 mg |
442.00€
0.10mg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
Ki67/MKI67 Monoclonal specifically detects Ki67/MKI67 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular), Knockout Validated.Spécification
Ki67/MKI67 | |
1.0 mg/mL | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay | |
Unconjugated | |
Rabbit | |
Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Mitotic Regulators, Neuroscience | |
PBS with 0.02% Sodium Azide | |
4288 | |
The immunogen for this KI67/MKI67 Antibody (1297A) was made using a synthetic peptide from the internal portion of Mouse KI67/MKI67, between amino acids 1850-1950 [UniProt# E9PVX6]. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
351 kDa |
1297A | |
Flow Cytometry 1 μg/mL, Immunohistochemistry 3-15 μg/mL, Immunocytochemistry/Immunofluorescence 1-10 μg/mL, Immunohistochemistry-Paraffin 3-15 μg/mL, Flow (Intracellular) 1 μg/mL, Knockout Validated | |
Monoclonal | |
Purified | |
RUO | |
Human | |
antigen identified by monoclonal Ki-67, antigen KI-67, Antigen Ki67, antiKi67, Ki67, Ki-67, Ki67 flow cytometry, Ki-67 flow cytometry, Ki67 ihc, Ki-67 ihc, Ki67 mouse, Ki-67 mouse, Ki67 western blot, Ki-67 western blot, KIA, Marker Of Proliferation Ki-67, MIB-, MIB-1, PPP1R105, Proliferation Marker Protein Ki-67, proliferation-related Ki-67 antigen, Protein Phosphatase 1, Regulatory Subunit 105 | |
MKI67 | |
IgG | |
Protein A or G purified | |
Ki67/MKI67 Antibody (1297A). |
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Ki67/MKI67 Antibody (1297A) - BSA Free, Novus Biologicals™