missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ki67/MKI67 Antibody (1297A) - BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody has been used in 2 publications
Marque: Bio-Techne NBP2-54791SS
148.00 EUR valable jusqu'au 2025-03-28
Utilisez le code promo "25306" pour bénéficier de cette offre.
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Ki67/MKI67 Monoclonal specifically detects Ki67/MKI67 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular), Knockout Validated.
Spécification
Ki67/MKI67 | |
Monoclonal | |
1.0 mg/mL | |
Flow Cytometry 1 μg/mL, Immunohistochemistry 3-15 μg/mL, Immunocytochemistry/Immunofluorescence 1-10 μg/mL, Immunohistochemistry-Paraffin 3-15 μg/mL, Flow (Intracellular) 1 μg/mL, Knockout Validated | |
antigen identified by monoclonal Ki-67, antigen KI-67, Antigen Ki67, antiKi67, Ki67, Ki-67, Ki67 flow cytometry, Ki-67 flow cytometry, Ki67 ihc, Ki-67 ihc, Ki67 mouse, Ki-67 mouse, Ki67 western blot, Ki-67 western blot, KIA, Marker Of Proliferation Ki-67, MIB-, MIB-1, PPP1R105, Proliferation Marker Protein Ki-67, proliferation-related Ki-67 antigen, Protein Phosphatase 1, Regulatory Subunit 105 | |
Rabbit | |
351 kDa | |
0.025 mg | |
Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Mitotic Regulators, Neuroscience | |
4288 | |
Human | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay | |
1297A | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
MKI67 | |
The immunogen for this KI67/MKI67 Antibody (1297A) was made using a synthetic peptide from the internal portion of Mouse KI67/MKI67, between amino acids 1850-1950 [UniProt# E9PVX6]. | |
Protein A or G purified | |
RUO | |
Primary | |
Ki67/MKI67 Antibody (1297A). | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Ki67/MKI67 Antibody (1297A) - BSA Free, Novus Biologicals™ > 0.025mg; Unlabeled
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu