Learn More
Novus Biologicals™ INSTA-Blot Mouse Tissues
Applications: SDS-Page
Marque: Novus Biologicals™ NBP2-30111-2Each
466.40 EUR valable jusqu'au 2025-03-28
Utilisez le code promo "25306" pour bénéficier de cette offre.
Description
- Screening proteins from various mouse tissues
- Contains 11 mouse tissue lysates (20μg total protein per lane) on 4-20% Tris-Glycine mini gel
- Prepared from resolution by SDS-PAGE at 125V for 1 hour
- Proteins are electro-blotted for 2 hours at 25V onto PVDF membrane
- Stained with amido black for visualization of proteins
Spécification
SDS-Page | |
2 Membrane | |
PVDF |