missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFNGR2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 10266-1-AP-150UL
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.00010kg
Description
This gene (IFNGR2) encodes the non-ligand-binding beta chain of the gamma interferon receptor. Human interferon-gamma receptor is a heterodimer of IFNGR1 and IFNGR2. Defects in IFNGR2 are a cause of mendelian susceptibility to mycobacterial disease (MSMD), also known as familial disseminated atypical mycobacterial infection. MSMD is a genetically heterogeneous disease with autosomal recessive, autosomal domit or X-linked inheritance.Spécification
IFNGR2 | |
Polyclonal | |
Unconjugated | |
IFNGR2 | |
AF 1, IFGR2, IFN gamma R2, IFN gamma receptor 2, IFNGR2, IFNGT1, Interferon gamma receptor 2, Interferon gamma transducer 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3460 | |
-20°C | |
Liquid |
Flow Cytometry, Immunoprecipitation, Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P38484 | |
IFNGR2 | |
IFNGR2 Fusion Protein Ag0317 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |