Learn More
R&D Systems™ Human TGF-beta 1 DuoSet ELISA
Human TGF-beta 1 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Marque: R&D Systems™ DY240-05
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The Human TGF-beta 1 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human TGF-beta 1.
Spécification
ELISA Development Kit | |
Human | |
31.20 to 2000 pg/mL | |
ELISA | |
CEDLAP, DPD1, latency-associated peptide, TGF beta1, TGFB, TGFB1, TGFbeta, TGFbeta 1, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1 | |
Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
1 Kit | |
Biotin | |
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules | |
7040 | |
5 Plates |
Correction du contenu d'un produit
Nom du produit
R&D Systems™ Human TGF-beta 1 DuoSet ELISA > Five 96-well plates
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu