missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human Osteoprotegerin (TNFRSF11B) Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Marque: Gibco™ 450-14-500UG
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The protein encoded by this gene is a member of the TNF-receptor superfamily. This protein is an osteoblast-secreted decoy receptor that functions as a negative regulator of bone resorption. This protein specifically binds to its ligand, osteoprotegerin ligand, both of which are key extracellular regulators of osteoclast development. Studies of the mouse counterpart also suggest that this protein and its ligand play a role in lymph-node organogenesis and vascular calcification. Alternatively spliced transcript variants of this gene have been reported, but their full length nature has not been determined.Spécification
O00300 | |
protein with no preservative | |
20 | |
500 μg | |
-20°C | |
<1 EU/μg | |
TNFRSF11B | |
Unconjugated | |
METFPPKYLH YDEETSHQLL CDKCPPGTYL KQHCTAKWKT VCAPCPDHYY TDSWHTSDEC LYCSPVCKEL QYVKQECNRT HNRVCECKEG RYLEIEFCLK HRSCPPGFGV VQAGTPERNT VCKRCPDGFF SNETSSKAPC RKHTNCSVFG LLLTQKGNAT HDNICSGNSE STQK | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
ELISA, Functional Assay, Western Blot | |
4982 | |
Human Osteoprotegerin (TNFRSF11B) | |
E. coli | |
RUO | |
LOW QUALITY PROTEIN: tumor necrosis factor receptor superfamily member 11 B; MGC29565; OCIF; Opg; osteoclastogenesis inhibitory factor; Osteoprotegerin; PDB5; soluble OPG; soluble Osteoprotegerin; sOPG; TNF receptor superfamily member 11 b; TNFRSF11B; TR1; tumor necrosis factor receptor superfamily member 11 B; tumor necrosis factor receptor superfamily, member 11 b; tumor necrosis factor receptor superfamily, member 11 b (osteoprotegerin) | |
Human | |
Recombinant | |
Lyophilized |