missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human IGFBP-3 Quantikine ELISA Kit
Human IGFBP-3 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
752.00€ - 4290.00€
Spécification
Plage de dosage | 0.78 - 50ng/mL |
---|---|
Sensibilité du dosage | 0.14ng/mL |
Conjugué | HRP |
Réactivité croisée | Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules |
Type de produit | Solid Phase Sandwich ELISA |
Code produit | Marque | Quantité | Suffisant pour | Prix | Quantité et disponibilité | ||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Suffisant pour | Prix | Quantité et disponibilité | ||||
18751974
|
R&D Systems™
DGB300 |
1 Plate | Sufficient materials to run ELISAs on at least one 96-well plate |
752.00€
1 kit |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
18761984
|
R&D Systems™
SGB300 |
6 Plates | Sufficient materials to run ELISAs on at least six 96-well plates |
4290.00€
1 kit |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
The Human IGFBP-3 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human IGFBP-3 in cell culture supernates (100 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul).Spécification
0.78 - 50ng/mL | |
HRP | |
Solid Phase Sandwich ELISA | |
Natural and Recombinant Human IGFBP-3 | |
ELISA | |
Cell Culture Supernates (100uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL) | |
acid stable subunit of the 140 K IGF complex, binding protein 29, binding protein 53, growth hormone-dependent binding protein, IBP-3, IBP3BP-53, IGF-binding protein 3, IGFBP3, IGFBP-3, insulin-like growth factor binding protein 3, insulin-like growth factor-binding protein 3 |
0.14ng/mL | |
Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules | |
Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum | |
Human | |
3486 | |
Store the unopened product at 2° to 8°C. Do not use past expiration date. |