missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human IGFBP-3 Quantikine ELISA Kit
Human IGFBP-3 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Marque: R&D Systems™ SGB300
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The Human IGFBP-3 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human IGFBP-3 in cell culture supernates (100 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul).
Spécification
0.78 - 50ng/mL | |
HRP | |
Solid Phase Sandwich ELISA | |
Natural and Recombinant Human IGFBP-3 | |
Human | |
3486 | |
Cell Culture Supernates (100uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL) | |
acid stable subunit of the 140 K IGF complex, binding protein 29, binding protein 53, growth hormone-dependent binding protein, IBP-3, IBP3BP-53, IGF-binding protein 3, IGFBP3, IGFBP-3, insulin-like growth factor binding protein 3, insulin-like growth factor-binding protein 3 |
0.14ng/mL | |
Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules | |
Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum | |
Sufficient materials to run ELISAs on at least six 96-well plates | |
ELISA | |
6 Plates | |
Store the unopened product at 2° to 8°C. Do not use past expiration date. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu