Learn More
Glypican 3 Antibody (GPC3/863), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NBP2-47762V
Description
Glypican 3 Monoclonal antibody specifically detects Glypican 3 in Human, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF
Spécification
Glypican 3 | |
Monoclonal | |
DyLight 405 | |
DGSX, glypican 3, glypican proteoglycan 3, glypican-3, GTR2-2, heparan sulphate proteoglycan, Intestinal protein OCI-5, MXR7, OCI5, OCI-5, secreted glypican-3, SGB, SGBS, SGBS1SDYS | |
Recombinant full-length human Glypican 3 protein (Uniprot: P51654) | |
0.1 mL | |
Cancer, Cell Biology, Cytoskeleton Markers, Signal Transduction, Stem Cells | |
2719 | |
Store at 4°C in the dark. | |
IgG1 κ |
Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence | |
GPC3/863 | |
50 mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.