Learn More
GATA3 Mouse anti-Human, Mouse, Clone: 1B11A8, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 66400-1-IG-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Trans-acting T-cell specific transcription factor GATA-3 is a member of GATA family of transcription factors that regulates development of multiple tissues. It is an important transcription factor in regulating human Th2 cell differentiation in vivo.Spécification
GATA3 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P23771, P23772 | |
Gata3 | |
GATA3 Fusion Protein Ag0174 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2a |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1B11A8 | |
Unconjugated | |
GATA3 | |
GATA binding factor 3, GATA binding protein 3, GATA3, HDR | |
Mouse | |
Protein A | |
RUO | |
14462, 2625 | |
-20°C | |