Learn More
Tocris Bioscience™ G 28UCM
FASN inhibitor
Marque: Tocris Bioscience™ 5446/10
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
G 28UCM is a FASN inhibitor. Inhibits breast cancer cell proliferation in vitro (IC50 values are 21 - 79 μM), including cell lines resistant to anti-HER2 therapies. Blocks HER2 signaling and induces apoptosis. Inhibits growth of breast cancer xenografts in mice without causing anorexia.Spécification
3-(3,4,5-Trihydroxybenzoyloxy)naphthalen-1-yl 3,4,5-trihydroxybenzoate | |
10 mg | |
C24H16O10 | |
>98% |
1094451-90-7 | |
Fatty Acid Synthase Inhibitors | |
464.38 |