Learn More
Labconco FreeZone 18L Console Freeze Dryer
FreeZone Console Freeze Dryers are designed for lyophilizing moderate to large sample loads or numerous small container batches. The console features a front viewing window, interior space for a vacuum pump, and accommodation for a full size tray dryer.
Marque: Labconco 701812190
Code nomenclature Nacres: NB.84
Informations supplémentaires : Poids : 308.39999kg
Description
- Stainless steel or PTFE coil holds 18 liters of ice before defrosting.
- The hp HCFC/CFC-free refrigeration system cools the collector
- Includes a stoppering tray dryer with heated and cooled shelves, vacuum release valve control, stoppering knob and more.
- Lyo-Works™ OS provides a real time display of collector temperature, vacuum level and optional End-Zone™ end point detection system
- Large, intuitive 5 in. full color, capacitive touchscreen display for easy operation.
- Stay Connected
- View Real-Time Sample Data
- Get Consistent, High Quality Results
- Make Challenging Samples Easy
Spécification
Console Freeze Dryer | |
0 | |
PTFE Coil and Collector Chamber | |
All except North America |
18 L | |
-50°C | |
Purge Valve | |
230 V |